siRNA Details
| Gene Symbol: DEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017418.2 | NP_059114.1 | 8 | 1091 | ||
| GenBank mRNA | AB022761.1 | - | 8 | 1094 | ||
| BC107103.1 | - | 8 | 1182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DEC1| Gene Symbol: DEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017418.2 | NP_059114.1 | 8 | 1091 | ||
| GenBank mRNA | AB022761.1 | - | 8 | 1094 | ||
| BC107103.1 | - | 8 | 1182 | |||