siRNA Details
| Gene Symbol: NUP54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017426.3 | NP_059122.2 | 3, 4 | 437 | ||
| XM_006714237.1 | XP_006714300.1 | 4, 5 | 414 | |||
| XM_011532033.2 | XP_011530335.1 | 3, 4 | 380 | |||
| XM_017008294.1 | XP_016863783.1 | 3, 4 | 824 | |||
| GenBank mRNA | AF157322.1 | - | 3, 4 | 318 | ||
| AK001517.1 | - | 3, 4 | 319 | |||
| AK222752.1 | - | 3, 4 | 324 | |||
| AK223352.1 | - | 3, 4 | 320 | |||
| AK315160.1 | - | 3, 4 | 317 | |||
| BC012559.1 | - | 3, 4 | 334 | |||
| DA577078.1 | - | 3, 4 | 437 | |||

NUP54
Genomic Map