siRNA Details
| Gene Symbol: NGEF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114090.1 | NP_001107562.1 | 10 | 1498 | ||
| NM_019850.2 | NP_062824.2 | 12 | 1870 | |||
| XM_011510923.2 | XP_011509225.1 | 12 | 1836 | |||
| XM_011510924.1 | XP_011509226.1 | 8 | 869 | |||
| XM_011510925.1 | XP_011509227.1 | 8 | 873 | |||
| GenBank mRNA | AK293203.1 | - | 8 | 870 | ||
| AK297390.1 | - | 10 | 1498 | |||
| AK298315.1 | - | 12 | 1822 | |||
| AK300953.1 | - | 13 | 1774 | |||
| BC031573.1 | - | 12 | 1857 | 5186132 | ||
| BX431262.1 | - | 7 | 743 | |||

NGEF
Genomic Map