siRNA Details
| Gene Symbol: ANKRD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129981.2 | NP_001123453.1 | 8 | 1372 | ||
| NM_001291218.1 | NP_001278147.1 | 9 | 1471 | |||
| NM_001291219.1 | NP_001278148.1 | 8 | 1372 | |||
| NM_020349.3 | NP_065082.2 | 9 | 1471 | |||
| GenBank mRNA | AB058599.1 | - | 9 | 1133 | ||
| AJ304805.1 | - | 9 | 1122 | |||
| AJ346356.1 | - | 3 | 407 | |||
| AJ583444.1 | - | 9 | 1460 | |||
| AK056990.1 | - | 9 | 1413 | |||
| BC020817.1 | - | 8 | 1034 | |||
| BC107759.1 | - | 9 | 1374 | 4096546 | ||

ANKRD2
Genomic Map