siRNA Details
| Gene Symbol: RTCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003729.3 | NP_003720.1 | 2, 3 | 372 | ||
| GenBank mRNA | AK292366.1 | - | 2, 3 | 321 | ||
| AK299793.1 | - | 2, 3 | 373 | |||
| AK307445.1 | - | 2, 3 | 293 | |||
| AU310497.1 | - | 2, 3 | 373 | |||
| DA361597.1 | - | 2, 3 | 314 | |||
| Y11651.1 | - | 2, 3 | 324 | |||

RTCA
Genomic Map