siRNA Details
| Gene Symbol: ZFYVE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281734.1 | NP_001268663.1 | 12 | 3842 | ||
| NM_001281735.1 | NP_001268664.1 | 9 | 2020 | |||
| NM_021260.3 | NP_067083.1 | 12 | 3884 | |||
| NM_178441.2 | NP_848535.1 | 9 | 2025 | |||
| XM_017021373.1 | XP_016876862.1 | 10 | 2549 | |||
| GenBank mRNA | AB046809.1 | - | 12 | 3709 | ||
| AF318319.1 | - | 9 | 2052 | |||
| BC014902.1 | - | 12 | 3761 | |||
| BC053520.1 | - | 12 | 3812 | 5493027 | ||
| CK005173.1 | - | 1 | 601 | |||

ZFYVE1
Genomic Map