siRNA Details
| Gene Symbol: NUDT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001248011.1 | NP_001234940.1 | 5, 6 | 1025 | ||
| NM_024047.4 | NP_076952.1 | 6, 7 | 1121 | |||
| NM_198038.2 | NP_932155.1 | 6, 7 | 763 | |||
| XM_011532032.2 | XP_011530334.1 | 6, 7 | 3300 | |||
| GenBank mRNA | AF265341.1 | - | 6, 7 | 762 | ||
| AF273028.1 | - | 6, 7 | 1085 | |||
| AK027295.1 | - | 6, 7 | 1111 | |||
| AK074845.1 | - | 6, 7 | 763 | |||
| AK075408.1 | - | 6, 7 | 1041 | |||
| AK308028.1 | - | 5, 6 | 792 | |||
| AY026252.1 | - | 6, 7 | 1116 | |||
| AY359123.1 | - | 6, 7 | 815 | |||
| BC000542.2 | - | 6, 7 | 881 | 3163944 | ||
| BI552154.1 | - | 766 | ||||

NUDT9
Genomic Map