siRNA Details
| Gene Symbol: CECR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163079.1 | NP_001156551.1 | 2 | 982 | ||
| NM_031890.3 | NP_114096.1 | 1 | 2046 | |||
| XM_011546124.2 | XP_011544426.1 | 1 | 2073 | |||
| XM_011546125.2 | XP_011544427.1 | 2 | 1392 | |||
| GenBank mRNA | AF307451.1 | - | 1 | 1976 | ||
| AK095609.1 | - | 2 | 976 | |||

CECR6
Genomic Map