siRNA Details
| Gene Symbol: CPXCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184771.1 | NP_001171700.1 | 3 | 902 | ||
| NM_033048.5 | NP_149037.4 | 3 | 940 | |||
| GenBank mRNA | AF284767.1 | - | 1 | 717 | ||
| AK098646.1 | - | 3 | 877 | |||
| AK313768.1 | - | 3 | 915 | |||
| BC027614.2 | - | 3 | 910 | 4838752 | ||

CPXCR1
Genomic Map