siRNA Details
Gene Symbol: C2CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015500.1 | NP_056315.1 | 7 | 1152 | ||
NM_199050.2 | NP_950251.1 | 6 | 651 | |||
XM_005261109.4 | XP_005261166.1 | 6 | 692 | |||
XM_011529522.1 | XP_011527824.1 | 7 | 1252 | |||
XM_011529523.2 | XP_011527825.1 | 8 | 1918 | |||
XM_011529524.2 | XP_011527826.1 | 7 | 1252 | |||
XM_017028315.1 | XP_016883804.1 | 4 | 388 | |||
GenBank mRNA | AB047784.1 | - | 7 | 1152 | ||
AB047785.1 | - | 7 | 1152 | |||
AK128550.1 | - | 4 | 491 | |||
AL833323.1 | - | 6 | 651 | |||
BC062323.1 | - | 6 | 633 | 5419622 |
