siRNA Details
| Gene Symbol: LRRC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031692.2 | NP_001026862.1 | 4 | 1495 | ||
| NM_005824.2 | NP_005815.2 | 5 | 1548 | |||
| XM_005250108.1 | XP_005250165.1 | 5 | 1911 | |||
| GenBank mRNA | AK225701.1 | - | 4 | 1389 | ||
| AK226021.1 | - | 4 | 1401 | |||
| AK290305.1 | - | 4 | 1386 | |||
| AY358889.1 | - | 4 | 1328 | |||
| BC027903.1 | - | 4 | 1373 | 5229061 | ||
| U32907.1 | - | 5 | 1440 | |||

LRRC17
Genomic Map