siRNA Details
| Gene Symbol: RAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308353.1 | NP_001295282.1 | 4 | 870 | ||
| NM_005855.3 | NP_005846.1 | 3 | 809 | |||
| XM_017003152.1 | XP_016858641.1 | 3 | 731 | |||
| XM_017003153.1 | XP_016858642.1 | 3 | 1039 | |||
| XM_017003154.1 | XP_016858643.1 | 3 | 816 | |||
| XM_017003156.1 | XP_016858645.1 | 3 | 757 | |||
| GenBank mRNA | AJ001014.1 | - | 3 | 709 | ||
| BC000548.2 | - | 3 | 701 | 3163522 | ||

RAMP1
Genomic Map