siRNA Details
| Gene Symbol: AKAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005858.3 | NP_005849.1 | 3, 4 | 161 | ||
| XM_011527624.2 | XP_011525926.1 | 3, 4 | 303 | |||
| XM_017026141.1 | XP_016881630.1 | 3, 4 | 306 | |||
| GenBank mRNA | BC037270.2 | - | 3, 4 | 151 | 4837309 | |
| DR005936.1 | - | 3, 4 | 164 | |||
| Y11997.1 | - | 3, 4 | 152 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: BC078448, BC060209, BC042602, BC125521, AK004801, and others.

AKAP8
Genomic Map