siRNA Details
| Gene Symbol: VAMP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003761.4 | NP_003752.2 | 2, 3 | 311 | ||
| GenBank mRNA | AF053233.1 | - | 2, 3 | 179 | ||
| BC001634.1 | - | 2, 3 | 208 | 3538672 | ||
| BM702226.1 | - | 2, 3 | 311 | |||
| BQ227618.1 | - | 2, 3 | 241 | |||
| BT006700.1 | - | 1, 2 | 155 | |||
| BX092149.1 | - | 2, 3 | 202 | |||
| CR456995.1 | - | 1, 2 | 155 | |||

VAMP8
Genomic Map