siRNA Details
| Gene Symbol: LANCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136574.1 | NP_001130046.1 | 10 | 2010 | ||
| NM_001136575.1 | NP_001130047.1 | 10 | 2005 | |||
| NM_006055.2 | NP_006046.1 | 10 | 2099 | |||
| XM_005246243.2 | XP_005246300.1 | 10 | 2120 | |||
| GenBank mRNA | Y11395.1 | - | 10 | 2039 | ||

LANCL1
Genomic Map