siRNA Details
| Gene Symbol: CRISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190986.1 | NP_001177915.1 | 8 | 900 | ||
| NM_006061.2 | NP_006052.2 | 8 | 888 | |||
| GenBank mRNA | AK223380.1 | - | 8 | 792 | ||
| AK292786.1 | - | 8 | 847 | |||
| AK303966.1 | - | 6 | 615 | |||
| BC069580.1 | - | 7 | 774 | 7262396 | ||
| BC069602.1 | - | 7 | 776 | 7262408 | ||
| X94323.1 | - | 8 | 801 | |||
| X95240.1 | - | 7 | 776 | |||

CRISP3
Genomic Map