siRNA Details
| Gene Symbol: CRISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190986.1 | NP_001177915.1 | 8 | 1835 | ||
| NM_006061.2 | NP_006052.2 | 8 | 1823 | |||
| GenBank mRNA | X94323.1 | - | 8 | 1736 | ||
| X95240.1 | - | 7 | 1711 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRISP3| Gene Symbol: CRISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190986.1 | NP_001177915.1 | 8 | 1835 | ||
| NM_006061.2 | NP_006052.2 | 8 | 1823 | |||
| GenBank mRNA | X94323.1 | - | 8 | 1736 | ||
| X95240.1 | - | 7 | 1711 | |||