siRNA Details
| Gene Symbol: PKDREJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006071.1 | NP_006062.1 | 1 | 2270 | ||
| GenBank mRNA | AF116458.1 | - | 1 | 2270 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PKDREJ| Gene Symbol: PKDREJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006071.1 | NP_006062.1 | 1 | 2270 | ||
| GenBank mRNA | AF116458.1 | - | 1 | 2270 | ||