siRNA Details
| Gene Symbol: IRF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006084.4 | NP_006075.3 | 9 | 1391 | ||
| GenBank mRNA | BC035716.2 | - | 8 | 1267 | 5588434 | |
| BI497082.1 | - | 1 | 130 | |||
| M87503.1 | - | 9 | 1298 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IRF9| Gene Symbol: IRF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006084.4 | NP_006075.3 | 9 | 1391 | ||
| GenBank mRNA | BC035716.2 | - | 8 | 1267 | 5588434 | |
| BI497082.1 | - | 1 | 130 | |||
| M87503.1 | - | 9 | 1298 | |||