siRNA Details
| Gene Symbol: TIMM17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006335.2 | NP_006326.1 | 6 | 726 | ||
| GenBank mRNA | AF106622.1 | - | 6 | 697 | ||
| AK023063.1 | - | 3 | 1900 | |||
| BC000294.2 | - | 6 | 696 | 2819455 | ||
| BC004439.1 | - | 6 | 696 | 2819455 | ||
| BC007106.1 | - | 6 | 707 | 4284092 | ||
| BC009784.1 | - | 6 | 718 | 3683125 | ||
| BC013823.1 | - | 6 | 685 | |||
| BC015098.1 | - | 6 | 724 | 3686369 | ||
| BC016817.1 | - | 4 | 581 | |||
| BC020833.1 | - | 6 | 704 | 4773149 | ||
| X97544.1 | - | 6 | 703 | |||

TIMM17A
Genomic Map