siRNA Details
| Gene Symbol: HMGN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006353.2 | NP_006344.1 | 2 | 462 | ||
| XM_017010171.1 | XP_016865660.1 | 2 | 521 | |||
| XM_017010172.1 | XP_016865661.1 | 3 | 585 | |||
| XM_017010173.1 | XP_016865662.1 | 4 | 623 | |||
| GenBank mRNA | AK311841.1 | - | 2 | 410 | ||
| AU143224.1 | - | 2 | 462 | |||
| BC001282.1 | - | 2 | 406 | 3452950 | ||
| BT009824.1 | - | 1 | 224 | |||
| U90549.1 | - | 1 | 915 | |||

HMGN4
Genomic Map