siRNA Details
| Gene Symbol: DCTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261412.1 | NP_001248341.1 | 12, 13 | 1206 | ||
| NM_001261413.1 | NP_001248342.1 | 11, 12 | 1200 | |||
| NM_006400.4 | NP_006391.1 | 11, 12 | 1215 | |||
| XM_011537761.2 | XP_011536063.1 | 12, 13 | 1167 | |||
| GenBank mRNA | AK055682.1 | - | 11, 12 | 1078 | ||
| AK096242.1 | - | 13, 14 | 1074 | |||
| AK098794.1 | - | 12, 13 | 1060 | |||
| AK222693.1 | - | 11, 12 | 1078 | |||
| AK292364.1 | - | 13, 14 | 1093 | |||
| AK314705.1 | - | 11, 12 | 1057 | |||
| AY189155.1 | - | 12, 13 | 938 | |||
| BC000718.2 | - | 11, 12 | 1064 | 3504109 | ||
| BC009468.1 | - | 11, 12 | 990 | 3613836 | ||
| BC014083.2 | - | 11, 12 | 1038 | 4303142 | ||
| FJ224315.1 | - | 11, 12 | 1025 | |||
| U50733.1 | - | 11, 12 | 1025 | |||

DCTN2
Genomic Map