siRNA Details
| Gene Symbol: MAB21L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006439.4 | NP_006430.1 | 1 | 2177 | ||
| GenBank mRNA | AF262032.1 | - | 1 | 1827 | ||
| AF370007.1 | - | 1 | 1078 | |||
| AK055665.1 | - | 1 | 1951 | |||
| BC009983.2 | - | 1 | 1675 | 4125321 | ||

MAB21L2
Genomic Map