siRNA Details
| Gene Symbol: CXCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006564.1 | NP_006555.1 | 2 | 641 | ||
| XM_005264809.2 | XP_005264866.1 | 2 | 715 | |||
| XM_011533290.2 | XP_011531592.1 | 2 | 1240 | |||
| XM_011533291.2 | XP_011531593.1 | 2 | 1191 | |||
| GenBank mRNA | AF007545.1 | - | 1 | 560 | ||
| AK313754.1 | - | 2 | 715 | |||
| BC033584.1 | - | 1 | 592 | 5225727 | ||
| JC209412.1 | - | 2 | 641 | |||
| U73529.1 | - | 1 | 590 | |||
| U73531.1 | - | 1 | 2022 | |||
| Y13248.1 | - | 2 | 641 | |||

CXCR6
Genomic Map