siRNA Details
| Gene Symbol: FCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK075140.1 | - | 5 | 438 | ||
| AK309540.1 | - | 6 | 467 | |||
| AY358081.1 | - | 5 | 427 | |||
| AY358283.1 | - | 5 | 464 | |||
| BC020731.1 | - | 5 | 438 | 4692272 | ||
| CR456808.1 | - | 6 | 465 | |||

FCN3
Genomic Map