siRNA Details
| Gene Symbol: RBBP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006606.2 | NP_006597.2 | 4, 5 | 408 | ||
| XM_005260652.2 | XP_005260709.1 | 3, 4 | 372 | |||
| GenBank mRNA | AF237576.1 | - | 4, 5 | 362 | ||
| AL832411.1 | - | 4, 5 | 356 | |||
| BC015938.1 | - | 4, 5 | 408 | 3922423 | ||

RBBP9
Genomic Map