siRNA Details
| Gene Symbol: CPLX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006651.3 | NP_006642.1 | 4 | 699 | ||
| XM_011513391.1 | XP_011511693.1 | 3 | 630 | |||
| GenBank mRNA | AB593095.1 | - | 4 | 676 | ||
| AK126578.1 | - | 5 | 571 | |||
| BC002471.2 | - | 4 | 699 | 3349779 | ||

CPLX1
Genomic Map