siRNA Details
| Gene Symbol: JTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006694.3 | NP_006685.1 | 5 | 1392 | ||
| GenBank mRNA | AF131797.1 | - | 5 | 973 | ||
| BC000996.2 | - | 5 | 1041 | 2901221 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

JTB| Gene Symbol: JTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006694.3 | NP_006685.1 | 5 | 1392 | ||
| GenBank mRNA | AF131797.1 | - | 5 | 973 | ||
| BC000996.2 | - | 5 | 1041 | 2901221 | ||