siRNA Details
| Gene Symbol: COPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006710.4 | NP_006701.1 | 8 | 1267 | ||
| NM_198189.2 | NP_937832.1 | 9 | 1332 | |||
| GenBank mRNA | AK289891.1 | - | 9 | 866 | ||
| BC003090.1 | - | 8 | 791 | 3503287 | ||
| BC036499.1 | - | 8 | 1401 | 5264527 | ||
| BC080617.1 | - | 8 | 763 | 6252628 | ||

COPS8
Genomic Map