siRNA Details
| Gene Symbol: C6orf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286474.1 | NP_001273403.1 | 4, 5 | 380 | ||
| NM_001286475.1 | NP_001273404.1 | 4, 5 | 380 | |||
| NM_006781.4 | NP_006772.3 | 4, 5 | 380 | |||
| XM_017010185.1 | XP_016865674.1 | 5, 6 | 405 | |||
| GenBank mRNA | AK225742.1 | - | 4, 5 | 371 | ||
| AK302176.1 | - | 4, 5 | 340 | |||
| AK310230.1 | - | 4, 5 | 369 | |||
| BC034774.1 | - | 4, 5 | 347 | 5169157 | ||
| DC399589.1 | - | 4, 5 | 381 | |||

C6orf10
Genomic Map