siRNA Details
| Gene Symbol: WDR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006784.2 | NP_006775.1 | 2 | 201 | ||
| GenBank mRNA | AF083217.1 | - | 2 | 182 | ||
| AK292438.1 | - | 2 | 196 | |||
| BP281064.1 | - | 2 | 201 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR3| Gene Symbol: WDR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006784.2 | NP_006775.1 | 2 | 201 | ||
| GenBank mRNA | AF083217.1 | - | 2 | 182 | ||
| AK292438.1 | - | 2 | 196 | |||
| BP281064.1 | - | 2 | 201 | |||