siRNA Details
| Gene Symbol: UGT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252274.2 | NP_001239203.2 | 2 | 315 | ||
| NM_001252275.2 | NP_001239204.2 | 2 | 315 | |||
| NM_001301239.1 | NP_001288168.1 | 1 | 253 | |||
| NM_006798.4 | NP_006789.3 | 2 | 315 | |||
| GenBank mRNA | AJ006054.1 | - | 1 | 262 | ||
| AK304249.1 | - | 2 | 315 | |||
| AK314209.1 | - | 2 | 313 | |||
| BC142640.1 | - | 1 | 316 | |||

UGT2A1
Genomic Map