siRNA Details
| Gene Symbol: HNRNPA0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006805.3 | NP_006796.1 | 1 | 820 | ||
| GenBank mRNA | BC001008.1 | - | 1 | 819 | 2964662 | |
| BC007271.1 | - | 1 | 548 | 3160840 | ||
| BC009284.2 | - | 1 | 592 | 4299262 | ||
| BC011972.1 | - | 1 | 811 | 3454140 | ||
| BC012980.2 | - | 1 | 819 | 3617551 | ||
| BC018800.1 | - | 1 | 817 | |||
| BC018949.2 | - | 1 | 819 | 4131137 | ||
| BC019271.1 | - | 1 | 799 | 2823669 | ||
| BC028976.1 | - | 1 | 820 | 4993025 | ||
| BC030249.1 | - | 1 | 820 | 5242912 | ||
| CR456986.1 | - | 1 | 536 | |||
| L10372.1 | - | 1 | 223 | |||
| U23803.1 | - | 1 | 800 | |||

HNRNPA0
Genomic Map