siRNA Details
| Gene Symbol: LMAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006816.2 | NP_006807.1 | 1 | 410 | ||
| GenBank mRNA | AK222609.1 | - | 1 | 177 | ||
| AK292099.1 | - | 1 | 187 | |||
| AK301607.1 | - | 2 | 94 | |||
| AK308027.1 | - | 1 | 321 | |||
| BC017263.2 | - | 1 | 190 | 4135210 | ||
| DC404202.1 | - | 1 | 410 | |||
| U10362.1 | - | 1 | 173 | |||

LMAN2
Genomic Map