siRNA Details
| Gene Symbol: CKAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006825.3 | NP_006816.2 | 2 | 2866 | ||
| GenBank mRNA | X69910.2 | - | 2 | 2864 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CKAP4| Gene Symbol: CKAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006825.3 | NP_006816.2 | 2 | 2866 | ||
| GenBank mRNA | X69910.2 | - | 2 | 2864 | ||