siRNA Details
| Gene Symbol: TDRKH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006862.3 | NP_006853.2 | 14 | 1987 | ||
| XM_017000126.1 | XP_016855615.1 | 14 | 1928 | |||
| XM_017000127.1 | XP_016855616.1 | 14 | 1957 | |||
| XM_017000130.1 | XP_016855619.1 | 14 | 1821 | |||
| GenBank mRNA | AF119121.2 | - | 14 | 1975 | ||
| BC032690.1 | - | 14 | 1894 | |||

TDRKH
Genomic Map