siRNA Details
| Gene Symbol: PIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006875.3 | NP_006866.2 | 6 | 1342 | ||
| GenBank mRNA | AK290931.1 | - | 6 | 1230 | ||
| BC018111.1 | - | 6 | 1213 | 3913552 | ||
| U77735.1 | - | 6 | 1226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIM2| Gene Symbol: PIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006875.3 | NP_006866.2 | 6 | 1342 | ||
| GenBank mRNA | AK290931.1 | - | 6 | 1230 | ||
| BC018111.1 | - | 6 | 1213 | 3913552 | ||
| U77735.1 | - | 6 | 1226 | |||