siRNA Details
| Gene Symbol: ZPBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159878.1 | NP_001153350.1 | 2 | 232 | ||
| NM_007009.2 | NP_008940.2 | 2 | 232 | |||
| XM_011515095.2 | XP_011513397.1 | 2 | 226 | |||
| XM_011515096.2 | XP_011513398.1 | 2 | 204 | |||
| XM_011515097.1 | XP_011513399.1 | 2 | 226 | |||
| XM_011515100.2 | XP_011513402.1 | 2 | 180 | |||
| XM_011515101.2 | XP_011513403.1 | 2 | 180 | |||
| XM_011515102.2 | XP_011513404.1 | 2 | 405 | |||
| XM_011515103.1 | XP_011513405.1 | 2 | 226 | |||
| XM_017011707.1 | XP_016867196.1 | 2 | 226 | |||
| GenBank mRNA | AK312799.1 | - | 2 | 204 | ||
| BC005223.1 | - | 2 | 191 | 3950015 | ||
| BT006822.1 | - | 2 | 162 | |||
| D17570.1 | - | 2 | 198 | |||

ZPBP
Genomic Map