siRNA Details
| Gene Symbol: CNTRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007018.4 | NP_008949.4 | 3 | 437 | ||
| XM_005251679.3 | XP_005251736.1 | 5 | 709 | |||
| XM_006716940.2 | XP_006717003.1 | 5 | 709 | |||
| XM_006716944.3 | XP_006717007.1 | 5 | 709 | |||
| XM_011518166.1 | XP_011516468.1 | 5 | 709 | |||
| XM_011518167.1 | XP_011516469.1 | 4 | 550 | |||
| XM_011518168.1 | XP_011516470.1 | 5 | 709 | |||
| XM_011518169.2 | XP_011516471.1 | 5 | 704 | |||
| XM_011518170.1 | XP_011516472.1 | 5 | 709 | |||
| XM_011518172.2 | XP_011516474.1 | 5 | 709 | |||
| XM_017014223.1 | XP_016869712.1 | 5 | 709 | |||
| XM_017014224.1 | XP_016869713.1 | 5 | 709 | |||
| XM_017014227.1 | XP_016869716.1 | 4 | 727 | |||
| XM_017014229.1 | XP_016869718.1 | 5 | 709 | |||
| XM_017014230.1 | XP_016869719.1 | 5 | 709 | |||
| GenBank mRNA | AF513978.1 | - | 3 | 406 | ||
| AK307387.1 | - | 5 | 714 | |||
| BC089415.1 | - | 5 | 666 | |||
| BC137284.1 | - | 3 | 430 | 9021281 | ||
| BC137286.1 | - | 3 | 429 | 9021283 | ||

CNTRL
Genomic Map