siRNA Details
| Gene Symbol: CYB561D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291284.1 | NP_001278213.1 | 4 | 791 | ||
| NM_007022.4 | NP_008953.1 | 4 | 857 | |||
| GenBank mRNA | AF040704.1 | - | 4 | 614 | ||
| AK291167.1 | - | 4 | 677 | |||
| BC047691.1 | - | 4 | 686 | 6060648 | ||
| BX641103.1 | - | 1 | 2698 | |||

CYB561D2
Genomic Map