siRNA Details
| Gene Symbol: UNC5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003728.3 | NP_003719.3 | 3 | 832 | ||
| XM_005263321.3 | XP_005263378.1 | 3 | 832 | |||
| XM_017008789.1 | XP_016864278.1 | 4 | 7249 | |||
| XM_017008790.1 | XP_016864279.1 | 3 | 477 | |||
| GenBank mRNA | AF055634.1 | - | 3 | 633 | ||
| AK290500.1 | - | 3 | 643 | |||
| AK308421.1 | - | 3 | 595 | |||
| BC041156.1 | - | 3 | 828 | 5208108 | ||

UNC5C
Genomic Map