siRNA Details
| Gene Symbol: WDHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008396.2 | NP_001008397.1 | 24 | 3153 | ||
| NM_007086.3 | NP_009017.1 | 25 | 3246 | |||
| XM_006720012.2 | XP_006720075.1 | 25 | 3238 | |||
| XM_011536373.2 | XP_011534675.1 | 24 | 3045 | |||
| XM_017020940.1 | XP_016876429.1 | 24 | 3151 | |||
| XM_017020941.1 | XP_016876430.1 | 22 | 2849 | |||
| GenBank mRNA | AJ006266.1 | - | 24 | 3195 | ||
| AK001538.1 | - | 13 | 1800 | |||
| AK001585.1 | - | 15 | 2169 | |||
| AK001620.1 | - | 8 | 973 | |||
| AK293005.1 | - | 25 | 3213 | |||
| BC063041.1 | - | 25 | 3222 | 6514641 | ||

WDHD1
Genomic Map