siRNA Details
Gene Symbol: CIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206999.1 | NP_001193928.1 | 42 | 5409 | ||
NM_007174.2 | NP_009105.1 | 41 | 5283 | |||
XM_006719206.2 | XP_006719269.1 | 42 | 5391 | |||
XM_011537783.1 | XP_011536085.1 | 43 | 5481 | |||
XM_011537784.1 | XP_011536086.1 | 43 | 5481 | |||
XM_011537785.1 | XP_011536087.1 | 43 | 5436 | |||
XM_011537787.1 | XP_011536089.1 | 41 | 5265 | |||
XM_011537788.1 | XP_011536090.1 | 41 | 5265 | |||
XM_011537789.1 | XP_011536091.1 | 35 | 4616 | |||
XM_011537790.2 | XP_011536092.1 | 34 | 4184 | |||
XM_011537791.2 | XP_011536093.1 | 34 | 5169 | |||
XM_011537792.2 | XP_011536094.1 | 33 | 5224 | |||
XM_017018735.1 | XP_016874224.1 | 42 | 5436 | |||
XM_017018736.1 | XP_016874225.1 | 43 | 5436 | |||
XM_017018737.1 | XP_016874226.1 | 42 | 5391 | |||
XM_017018738.1 | XP_016874227.1 | 32 | 4725 | |||
XM_017018739.1 | XP_016874228.1 | 32 | 5500 | |||
GenBank mRNA | AB023166.2 | - | 33 | 5224 | ||
AY257469.1 | - | 41 | 5280 | |||
AY681966.1 | - | 42 | 5409 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
