siRNA Details
| Gene Symbol: C14orf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007176.3 | NP_009107.1 | 5 | 854 | ||
| GenBank mRNA | AF134159.2 | - | 5 | 474 | ||
| AF136971.1 | - | 5 | 514 | |||
| AF161406.1 | - | 5 | 514 | |||
| AK311764.1 | - | 5 | 494 | |||
| AL136658.1 | - | 5 | 512 | |||
| BC002444.2 | - | 5 | 521 | 3347242 | ||
| BX248022.1 | - | 5 | 848 | |||
| CR533443.1 | - | 4 | 402 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_021446, BC004591, AK004480, AF270646, AK002308, and others.

C14orf1
Genomic Map