siRNA Details
| Gene Symbol: WBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007187.3 | NP_009118.1 | 5 | 512 | ||
| XM_005266245.2 | XP_005266302.1 | 5 | 529 | |||
| XM_017020380.1 | XP_016875869.1 | 5 | 489 | |||
| GenBank mRNA | AF071185.1 | - | 5 | 548 | ||
| AK000979.1 | - | 5 | 564 | |||
| AK001557.1 | - | 5 | 547 | |||
| AK297536.1 | - | 4 | 450 | |||
| AK314525.1 | - | 5 | 549 | |||
| BC104879.1 | - | 5 | 456 | 8143882 | ||
| BC108310.1 | - | 5 | 513 | 6014541 | ||

WBP4
Genomic Map