siRNA Details
| Gene Symbol: WBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007187.3 | NP_009118.1 | 10 | 1235 | ||
| XM_005266245.2 | XP_005266302.1 | 10 | 1252 | |||
| XM_017020380.1 | XP_016875869.1 | 9 | 1165 | |||
| GenBank mRNA | AF071185.1 | - | 10 | 1271 | ||
| AK000979.1 | - | 10 | 1288 | |||
| AK001557.1 | - | 10 | 1270 | |||
| AK297536.1 | - | 9 | 1173 | |||
| BC104879.1 | - | 10 | 1179 | 8143882 | ||
| BC108310.1 | - | 10 | 1236 | 6014541 | ||

WBP4
Genomic Map