siRNA Details
| Gene Symbol: SEC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007214.4 | NP_009145.1 | 21 | 2660 | ||
| XM_011535399.2 | XP_011533701.1 | 21 | 2409 | |||
| XM_017010218.1 | XP_016865707.1 | 21 | 2489 | |||
| GenBank mRNA | AF119883.1 | - | 1 | 246 | ||
| AJ420532.1 | - | 3 | 1201 | |||
| AK027374.1 | - | 17 | 1933 | |||
| AK074835.1 | - | 21 | 2476 | |||
| AL137338.1 | - | 18 | 2007 | |||
| BC047221.1 | - | 21 | 2529 | 5277083 | ||
| BC048287.1 | - | 21 | 2516 | 5212007 | ||

SEC63
Genomic Map