siRNA Details
| Gene Symbol: RNF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007282.4 | NP_009213.1 | 9 | 1416 | ||
| NM_183381.2 | NP_899237.1 | 8 | 1314 | |||
| XM_005247092.3 | XP_005247149.1 | 8 | 1273 | |||
| XM_011512373.2 | XP_011510675.1 | 10 | 1421 | |||
| XM_011512374.2 | XP_011510676.1 | 9 | 1319 | |||
| XM_011512376.2 | XP_011510678.1 | 8 | 724 | |||
| XM_017005654.1 | XP_016861143.1 | 9 | 1495 | |||
| XM_017005655.1 | XP_016861144.1 | 8 | 1396 | |||
| XM_017005656.1 | XP_016861145.1 | 9 | 1375 | |||
| XM_017005657.1 | XP_016861146.1 | 9 | 1508 | |||
| XM_017005658.1 | XP_016861147.1 | 8 | 1406 | |||
| XM_017005659.1 | XP_016861148.1 | 7 | 1143 | |||
| XM_017005660.1 | XP_016861149.1 | 7 | 1133 | |||
| XM_017005661.1 | XP_016861150.1 | 2 | 498 | |||
| GenBank mRNA | AF037204.1 | - | 8 | 783 | ||
| AF070558.1 | - | 8 | 774 | |||
| AK090638.1 | - | 8 | 1294 | |||
| AK090771.1 | - | 5 | 403 | |||
| AK313304.1 | - | 8 | 798 | |||
| BC009781.1 | - | 8 | 791 | 3683407 | ||
| BC009803.1 | - | 9 | 894 | 4274992 | ||
| BC017878.1 | - | 8 | 756 | |||
| BG546092.1 | - | 6 | 587 | |||
| BM556246.1 | - | 7 | 669 | |||
| CB109037.1 | - | 2 | 141 | |||
| CR456804.1 | - | 7 | 632 | |||

RNF13
Genomic Map