siRNA Details
| Gene Symbol: KLRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007360.3 | NP_031386.2 | 8 | 977 | ||
| GenBank mRNA | AK226161.1 | - | 6 | 1911 | ||
| BC039836.1 | - | 8 | 941 | 6067176 | ||
| Gene Symbol: KLRC4-KLRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199805.1 | NP_001186734.1 | 13 | 1335 | ||
| GenBank mRNA | AK292020.1 | - | 13 | 1448 | ||
| X54870.1 | - | 10 | 1138 | |||

KLRK1
Genomic Map