siRNA Details
| Gene Symbol: CLCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012128.3 | NP_036260.2 | 4 | 568 | ||
| XM_011541015.2 | XP_011539317.1 | 4 | 387 | |||
| GenBank mRNA | AF127035.1 | - | 4 | 552 | ||
| AK000072.1 | - | 4 | 566 | |||
| AK314430.1 | - | 4 | 564 | |||
| AY358470.1 | - | 4 | 548 | |||
| BC113687.1 | - | 4 | 563 | 8322739 | ||
| BC113689.1 | - | 4 | 562 | 8322741 | ||

CLCA4
Genomic Map